Assay Details
Target Gene Details
Entrez Gene ID: | 22072 |
Gene Name: | protease, serine 2 |
Gene Aliases: |
TRY8, TRYP, Ta, Tesp4, Try2 |
Location: |
Chr.6:41521776-41525079 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss2 | NM_009430.2 | NP_033456.1 | ||
| AK003082.1 | BAB22554.1 | |||
| X04574.1 | CAA28243.1 |
More Information
Set Membership: |
|

Genomic Map