Assay Details
Target Gene Details
Entrez Gene ID: | 19362 |
Gene Name: | RAD51 associated protein 1 |
Gene Aliases: |
2510006L10Rik, RAB22 |
Location: |
Chr.6:126923415-126939621 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad51ap1 | NM_009013.3 | 8 | 837 | NP_033039.2 |
| AK010920.1 | 6 | 836 | ||
| AK011379.1 | 8 | 838 | BAB27579.1 | |
| AK077829.1 | 8 | 867 | BAC37025.1 | |
| AK079525.1 | 8 | 859 | BAC37671.1 | |
| BC003738.1 | 8 | 851 | AAH03738.1 | |
| U93583.1 | 8 | 835 | AAB91541.1 |

Genomic Map