Assay Details
Target Gene Details
Entrez Gene ID: | 68240 |
Gene Name: | replication protein A3 |
Gene Aliases: |
14kDa, C330026P08Rik |
Location: |
Chr.6:8255936-8259141 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpa3 | NM_026632.4 | NP_080908.1 | ||
| AK005285.1 | BAB23932.1 | |||
| AK009916.1 | BAB26583.1 | |||
| AK021228.1 | BAB32338.1 | |||
| AK088697.1 | 1 | 904 | ||
| BC028489.1 | AAH28489.1 | |||
| BY057686.1 |

Genomic Map