Assay Details
Target Gene Details
Entrez Gene ID: | 330267 |
Gene Name: | thrombospondin, type I, domain containing 7A |
Gene Aliases: |
Gm837 |
Location: |
Chr.6:12311608-12749253 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thsd7a | NM_001164805.1 | 25 | 4820 | NP_001158277.1 |
| AK133324.1 | 20 | 2900 | ||
| AK173072.1 | 25 | 4591 | BAD32350.1 |
More Information
Set Membership: |
|

Genomic Map