Assay Details
Target Gene Details
Entrez Gene ID: | 320938 |
Gene Name: | transportin 3 |
Gene Aliases: |
5730544L10Rik, C430013M08Rik, C81142, D6Ertd313e, Trn-SR, mKIAA4133 |
Location: |
Chr.6:29540827-29610110 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnpo3 | NM_177296.4 | 22 | 2886 | NP_796270.2 |
| AK011912.1 | 3 | 191 | BAB27912.1 | |
| AK077714.1 | 6 | 734 | BAC36974.1 | |
| AK220184.1 | 15 | 1789 | BAD90369.1 | |
| BC052756.1 | 11 | 1207 | AAH52756.1 | |
| BC064646.1 | 22 | 2886 | AAH64646.1 | |
| BC075678.1 | 22 | 3099 | AAH75678.1 |
More Information
Set Membership: |
|

Genomic Map