Assay Details
Target Gene Details
Entrez Gene ID: | 100503991 |
Gene Name: | acid phosphatase, testicular |
Gene Aliases: |
EG546967, Gm1432 |
Location: |
Chr.7:44251761-44261396 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acpt | NM_001195034.1 | NP_001181963.1 |
More Information
Set Membership: |
|

Genomic Map