Assay Details
Target Gene Details
Entrez Gene ID: | 233649 |
Gene Name: | cyclic nucleotide gated channel alpha 4 |
Gene Aliases: |
- |
Location: |
Chr.7:105400377-105409313 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnga4 | NM_001033317.3 | 1 | 53 | NP_001028489.1 |
| AK136723.1 | 1 | 55 | BAE23107.1 | |
| BC107347.1 | AAI07348.1 | |||
| BC107348.2 |
More Information
Set Membership: |
|

Genomic Map