Assay Details
Target Gene Details
Entrez Gene ID: | 233649 |
Gene Name: | cyclic nucleotide gated channel alpha 4 |
Gene Aliases: |
- |
Location: |
Chr.7:105400377-105409313 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnga4 | NM_001033317.3 | NP_001028489.1 | ||
| AK136723.1 | BAE23107.1 | |||
| BC107347.1 | AAI07348.1 | |||
| BC107348.2 | 2 | 428 |
More Information
Set Membership: |
|

Genomic Map