Assay Details
Target Gene Details
Entrez Gene ID: | 29861 |
Gene Name: | D4, zinc and double PHD fingers family 1 |
Gene Aliases: |
Neud4 |
Location: |
Chr.7:29303959-29317592 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpf1 | NM_013874.2 | 4 | 438 | NP_038902.1 |
| AK142419.1 | 4 | 417 | ||
| BC052348.1 | 4 | 467 | AAH52348.1 | |
| U48238.1 | 4 | 437 | AAF21455.1 |
More Information
Set Membership: |
|

Genomic Map