Assay Details
Target Gene Details
Entrez Gene ID: | 80794 |
Gene Name: | Casitas B-lineage lymphoma c |
Gene Aliases: |
2310076I21Rik, 2310079L19Rik, Cbl3 |
Location: |
Chr.7:19779718-19796809 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cblc | NM_001161844.1 | NP_001155316.1 | ||
| NM_023224.5 | NP_075713.3 | |||
| AK010228.1 | BAB26782.1 | |||
| BC046337.1 | AAH46337.1 | |||
| BC111037.1 | AAI11038.1 | |||
| BQ031029.1 |
More Information
Set Membership: |
|

Genomic Map