Assay Details
Target Gene Details
Entrez Gene ID: | 13170 |
Gene Name: | D site albumin promoter binding protein |
Gene Aliases: |
- |
Location: |
Chr.7:45705095-45710203 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbp | NM_016974.3 | NP_058670.2 | ||
| AK140243.1 | BAE24294.1 | |||
| AK181839.1 | ||||
| BC018323.1 | AAH18323.1 | |||
| BC084676.1 | AAH84676.1 | |||
| BP770661.1 |
More Information
Set Membership: |
|

Genomic Map