Assay Details
Target Gene Details
Entrez Gene ID: | 67605 |
Gene Name: | AKT1 substrate 1 (proline-rich) |
Gene Aliases: |
1110012J22Rik, AI227026, Lobe, Lobel, PRAS40 |
Location: |
Chr.7:44849133-44855421 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akt1s1 | NM_001290695.1 | NP_001277624.1 | ||
| NM_026270.4 | NP_080546.1 | |||
| AA231178.1 | ||||
| AK003638.1 | BAB22905.1 | |||
| AK030571.1 | ||||
| AK141771.1 | BAE24829.1 | |||
| AK154110.1 | BAE32383.1 | |||
| BB618044.1 | ||||
| BC132372.1 | ||||
| BC132374.1 | ||||
| BY103589.1 | ||||
| W71279.1 |

Genomic Map