Assay Details
Target Gene Details
Entrez Gene ID: | 58804 |
Gene Name: | CDC42 effector protein (Rho GTPase binding) 5 |
Gene Aliases: |
1700027J19Rik, 2010007O02Rik, Borg3, C85526, CEP5 |
Location: |
Chr.7:4151260-4164737 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42ep5 | NM_021454.3 | 1 | 165 | NP_067429.1 |
| AK008154.1 | 1 | 165 | BAB25498.1 | |
| BC006758.1 | AAH06758.1 | |||
| BC103774.1 | 1 | 176 | AAI03775.1 |

Genomic Map