Assay Details
Target Gene Details
Entrez Gene ID: | 12721 |
Gene Name: | coronin, actin binding protein 1A |
Gene Aliases: |
Clabp, Lmb3, TACO, p57 |
Location: |
Chr.7:126699774-126704816 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coro1a | NM_001301374.1 | NP_001288303.1 | ||
| NM_009898.3 | NP_034028.1 | |||
| AF047388.1 | AAD31082.1 | |||
| AF143955.1 | AAD32703.1 | |||
| AF495468.1 | AAM18514.1 | |||
| AK040885.1 | ||||
| AK075799.1 | ||||
| AK146290.1 | BAE27047.1 | |||
| AK150962.1 | BAE29993.1 | |||
| AK151755.1 | BAE30663.1 | |||
| AK155527.1 | BAE33310.1 | |||
| AK155860.1 | BAE33465.1 | |||
| AK165296.1 | BAE38125.1 | |||
| AK172447.1 | BAE43009.1 | |||
| BC002136.1 | AAH02136.1 | |||
| BC053398.1 | AAH53398.1 |
More Information
Set Membership: |
|

Genomic Map