Assay Details
Target Gene Details
Entrez Gene ID: | 59288 |
Gene Name: | dynactin 5 |
Gene Aliases: |
4930427E12Rik, C78178, b2b315Clo |
Location: |
Chr.7:122133041-122149044 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dctn5 | NM_021608.3 | NP_067621.3 | ||
| AF190795.1 | AAF05615.1 | |||
| AK010238.1 | BAB26789.1 | |||
| AK051957.1 | BAC34820.1 | |||
| AK076100.1 | BAC36183.1 | |||
| AK076738.1 | BAC36460.1 | |||
| AK145958.1 | ||||
| AK146123.1 | BAE26917.1 | |||
| AK151193.1 | BAE30190.1 | |||
| AK151876.1 | BAE30763.1 | |||
| AK168983.1 | BAE40783.1 | |||
| BC005671.1 | AAH05671.1 | |||
| BC010834.1 | AAH10834.1 |
More Information
Set Membership: |
|

Genomic Map