Assay Details
Target Gene Details
Entrez Gene ID: | 101744 |
Gene Name: | iron/zinc purple acid phospatase-like |
Gene Aliases: |
AU017076, C330005M16Rik |
Location: |
Chr.7:28607634-28631361 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Papl | NM_175319.4 | 4 | 719 | NP_780528.1 |
| AK049131.1 | 4 | 720 | BAC33559.1 | |
| BC094908.1 | 4 | 615 | AAH94908.1 | |
| BC132375.1 | 4 | 706 | ||
| BC138307.1 | 4 | 705 |
More Information
Set Membership: |
|

Genomic Map