Assay Details
Target Gene Details
Entrez Gene ID: | 12862 |
Gene Name: | cytochrome c oxidase subunit VIa polypeptide 2 |
Gene Aliases: |
CoxVIaH, VIaH |
Location: |
Chr.7:128205435-128206366 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox6a2 | NM_009943.2 | 2 | 215 | NP_034073.2 |
| AK002506.1 | 2 | 166 | BAB22151.1 | |
| BC028514.1 | 2 | 186 | AAH28514.1 | |
| BF472154.1 | 2 | 98 | ||
| BU842968.1 | 2 | 215 | ||
| U08439.1 | 2 | 201 | AAA17835.1 |
More Information
Set Membership: |
|

Genomic Map