Assay Details
Target Gene Details
Entrez Gene ID: | 67379 |
Gene Name: | death effector domain-containing DNA binding protein 2 |
Gene Aliases: |
2410050E11Rik, FLAME-3 |
Location: |
Chr.7:25202838-25219873 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dedd2 | NM_207677.3 | NP_997560.3 | ||
| AF457576.1 | AAM10836.1 | |||
| AF543541.1 | AAN33179.1 | |||
| AK010701.1 | ||||
| AK011713.1 | ||||
| BC092250.1 | AAH92250.1 | |||
| CX201206.1 |

Genomic Map