Assay Details
Target Gene Details
Entrez Gene ID: | 12865 |
Gene Name: | cytochrome c oxidase subunit VIIa 1 |
Gene Aliases: |
COX7A, COX7AH, COX7AM |
Location: |
Chr.7:30184171-30186030 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox7a1 | NM_009944.3 | 3 | 201 | NP_034074.1 |
| AF037370.1 | 3 | 164 | AAC23485.1 | |
| AK079950.1 | 3 | 202 | BAC37791.1 | |
| BC060974.1 | 3 | 179 | AAH60974.1 |
More Information
Set Membership: |
|

Genomic Map