Assay Details
Target Gene Details
Entrez Gene ID: | 13088 |
Gene Name: | cytochrome P450, family 2, subfamily b, polypeptide 10 |
Gene Aliases: |
Cyp2b, Cyp2b20, p16 |
Location: |
Chr.7:25897658-25926624 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2b10 | NM_009999.4 | 4 | 631 | NP_034129.1 |
| AF128849.1 | 4 | 613 | AAD28466.1 | |
| AF129405.1 | 3 | 427 | ||
| AI304065.1 | 4 | 585 | ||
| AK028103.1 | 4 | 631 | BAC25748.1 | |
| BC060973.1 | 4 | 625 | AAH60973.1 | |
| M21856.1 | 4 | 613 | AAA40425.1 | |
| X99715.1 | 4 | 609 | CAA68051.1 |
More Information
Set Membership: |
|

Genomic Map