Assay Details
Target Gene Details
Entrez Gene ID: | 232946 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 3 |
Gene Aliases: |
BC043666, Blos3, E230011O18, rp |
Location: |
Chr.7:19505804-19508371 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bloc1s3 | NM_177692.3 | NP_808360.2 | ||
| AK054006.1 | BAC35618.1 | |||
| BC043666.1 | AAH43666.1 |
Target Gene Details
Entrez Gene ID: | 67091 |
Gene Name: | trafficking protein particle complex 6A |
Gene Aliases: |
1810073E21Rik, 4930519D19Rik, AI480686, TRS33, mhyp |
Location: |
Chr.7:19507406-19516145 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc6a |
More Information
Set Membership: |
|

Genomic Map