Assay Details
Target Gene Details
Entrez Gene ID: | 54378 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 6 |
Gene Aliases: |
2310033H20Rik, AW050150 |
Location: |
Chr.7:3423580-3435667 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng6 | NM_133183.1 | NP_573446.1 | ||
| AF361348.1 | AAL50043.1 | |||
| BC115456.1 | ||||
| BC115457.1 |
More Information
Set Membership: |
|

Genomic Map