Assay Details
Target Gene Details
Entrez Gene ID: | 81905 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 8 |
Gene Aliases: |
mir-935 |
Location: |
Chr.7:3394117-3415605 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng8 | NM_133190.1 | 3 | 536 | NP_573453.1 |
| AF361350.1 | 3 | 536 | AAL50045.1 |
More Information
Set Membership: |
|

Genomic Map