Assay Details
Target Gene Details
Entrez Gene ID: | 243881 |
Gene Name: | cytochrome P450, family 2, subfamily b, polypeptide 23 |
Gene Aliases: |
EG243881 |
Location: |
Chr.7:26665227-26690372 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2b23 | NM_001081148.1 | 1 | 48 | NP_001074617.1 |
More Information
Set Membership: |
|

Genomic Map