Assay Details
Target Gene Details
Entrez Gene ID: | 76686 |
Gene Name: | CAP-GLY domain containing linker protein 3 |
Gene Aliases: |
1500005P14Rik, AI844915, Clipr-59 |
Location: |
Chr.7:30291688-30308367 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clip3 | NM_001081114.1 | NP_001074583.1 | ||
| BC138413.1 |
More Information
Set Membership: |
|

Genomic Map