Assay Details
Target Gene Details
Entrez Gene ID: | 13106 |
Gene Name: | cytochrome P450, family 2, subfamily e, polypeptide 1 |
Gene Aliases: |
Cyp2e |
Location: |
Chr.7:140763832-140774981 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2e1 | NM_021282.2 | 6 | 916 | NP_067257.1 |
| BC013451.1 | 6 | 916 | AAH13451.1 | |
| DQ838976.1 | ||||
| DQ838977.1 | ||||
| L11650.1 | 6 | 1009 | AAA39879.1 | |
| X62595.1 | 6 | 901 | CAA44481.1 |
More Information
Set Membership: |
|

Genomic Map