Assay Details
Target Gene Details
Entrez Gene ID: | 233913 |
Gene Name: | cDNA sequence BC017158 |
Gene Aliases: |
- |
Location: |
Chr.7:128271379-128298165 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC017158 | NM_145590.2 | NP_663565.2 | ||
| AK148236.1 | BAE28428.1 | |||
| AK149645.1 | BAE29003.1 | |||
| AK154566.1 | ||||
| AK164488.1 | ||||
| AK165287.1 | ||||
| AK170611.1 | BAE41911.1 | |||
| AK186460.1 | ||||
| BC017158.1 | AAH17158.1 |

Genomic Map