Assay Details
Target Gene Details
Entrez Gene ID: | 12315 |
Gene Name: | calmodulin 3 |
Gene Aliases: |
CaMA, R75142 |
Location: |
Chr.7:16915379-16924032 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calm3 | NM_007590.3 | NP_031616.1 | ||
| AK151610.1 | BAE30549.1 | |||
| AK152754.1 | BAE31469.1 | |||
| AK153179.1 | BAE31782.1 | |||
| AK165568.1 | ||||
| AK169630.1 | ||||
| BC050926.1 | AAH50926.1 | |||
| BE853985.1 | ||||
| M19380.1 | AAA66181.1 |
More Information
Set Membership: |
|

Genomic Map