Assay Details
Target Gene Details
Entrez Gene ID: | 12752 |
Gene Name: | ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease) |
Gene Aliases: |
AI323623 |
Location: |
Chr.7:126571400-126585817 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cln3 | NM_001146311.1 | NP_001139783.1 | ||
| NM_009907.3 | NP_034037.3 | |||
| AK078976.1 | BAC37491.1 | |||
| AK134036.1 | ||||
| AK184160.1 | ||||
| AK189026.1 | ||||
| AK217815.1 | 3 | 213 | ||
| BC058753.1 | AAH58753.1 | |||
| BC060306.1 | AAH60306.1 | |||
| BC080759.1 | AAH80759.1 | |||
| U47106.1 | AAC52957.1 | |||
| U68064.1 | AAB07595.1 | |||
| U92811.1 | AAB69983.1 |
More Information
Set Membership: |
|

Genomic Map