Assay Details
Target Gene Details
Entrez Gene ID: | 13108 |
Gene Name: | cytochrome P450, family 2, subfamily g, polypeptide 1 |
Gene Aliases: |
- |
Location: |
Chr.7:26808927-26821197 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2g1 | NM_013809.1 | 2 | 262 | NP_038837.1 |
| AK014160.1 | 2 | 282 | ||
| BC058222.1 | 2 | 267 | AAH58222.1 | |
| L81171.1 | 2 | 262 | AAD45178.1 |
More Information
Set Membership: |
|

Genomic Map