Assay Details
Target Gene Details
Entrez Gene ID: | 232910 |
Gene Name: | adaptor-related protein complex 2, sigma 1 subunit |
Gene Aliases: |
AI043088 |
Location: |
Chr.7:16738444-16749290 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap2s1 | NM_198613.2 | NP_941015.2 | ||
| AK146584.1 | BAE27279.1 | |||
| AK186335.1 | ||||
| AK206960.1 | ||||
| BC052499.1 | AAH52499.1 |
More Information
Set Membership: |
|

Genomic Map