Assay Details
Target Gene Details
Entrez Gene ID: | 13019 |
Gene Name: | cardiotrophin 1 |
Gene Aliases: |
CT-1 |
Location: |
Chr.7:127712704-127718188 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctf1 | NM_007795.1 | NP_031821.1 | ||
| BC024381.1 | AAH24381.1 | |||
| U18366.1 | AAC52173.1 |

Genomic Map