Assay Details
Target Gene Details
Entrez Gene ID: | 384724 |
Gene Name: | cytochrome P450, family 2, subfamily t, polypeptide 4 |
Gene Aliases: |
Gm1448, Gm1450 |
Location: |
Chr.7:27153714-27158973 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2t4 | NM_001100184.1 | 8 | 1052 | NP_001093654.1 |
More Information
Set Membership: |
|

Genomic Map