Assay Details
Target Gene Details
Entrez Gene ID: | 13389 |
Gene Name: | delta-like 3 (Drosophila) |
Gene Aliases: |
pu, pudgy |
Location: |
Chr.7:28293555-28301785 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dll3 | NM_007866.2 | NP_031892.2 | ||
| AB013440.1 | BAA33716.1 | |||
| AK144282.1 | BAE25812.1 | |||
| BB868475.1 | ||||
| BC052002.1 | AAH52002.1 |

Genomic Map