Assay Details
Target Gene Details
Entrez Gene ID: | 232975 |
Gene Name: | ATPase, Na+/K+ transporting, alpha 3 polypeptide |
Gene Aliases: |
Atpa-2 |
Location: |
Chr.7:24978167-25005937 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp1a3 | NM_001290469.1 | NP_001277398.1 | ||
| BC020177.1 | AAH20177.1 | |||
| BC027000.1 | AAH27000.1 | |||
| BC027114.1 | AAH27114.1 | |||
| BC034645.1 | AAH34645.1 | |||
| BC037206.1 | AAH37206.1 | |||
| BC042894.1 | AAH42894.1 |
Target Gene Details
Entrez Gene ID: | 102465634 |
Gene Name: | microRNA 7047 |
Gene Aliases: |
Gm27270, mmu-mir-7047 |
Location: |
Chr.7:24987604-24987667 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7047 | NR_106014.1 |
More Information
Set Membership: |
|

Genomic Map