Assay Details
Target Gene Details
Entrez Gene ID: | 12476 |
Gene Name: | CD151 antigen |
Gene Aliases: |
PETA-3, SFA-1, Tspan24 |
Location: |
Chr.7:141467362-141471481 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd151 | NM_001111049.1 | NP_001104519.1 | ||
| NM_009842.3 | 1 | 96 | NP_033972.2 | |
| AK178787.1 | ||||
| BQ896459.1 | 1 | 114 | ||
| D89290.1 | 1 | 65 | BAA22447.1 | |
| DV063036.1 |

Genomic Map