Assay Details
Target Gene Details
Entrez Gene ID: | 12476 |
Gene Name: | CD151 antigen |
Gene Aliases: |
PETA-3, SFA-1, Tspan24 |
Location: |
Chr.7:141467362-141471481 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd151 | NM_001111049.1 | 4 | 468 | NP_001104519.1 |
| NM_001111050.1 | 3 | 343 | NP_001104520.1 | |
| NM_009842.3 | 4 | 586 | NP_033972.2 | |
| BC012236.1 | 3 | 265 | AAH12236.1 | |
| BQ896459.1 | 4 | 604 | ||
| CJ066904.1 | 3 | 343 | ||
| D89290.1 | 4 | 555 | BAA22447.1 | |
| DV063036.1 | 4 | 495 | ||
| U89772.1 | 3 | 310 | AAC25976.1 |

Genomic Map