Assay Details
Target Gene Details
Entrez Gene ID: | 19158 |
Gene Name: | cytohesin 2 |
Gene Aliases: |
ARNO, CLM2, Pscd2 |
Location: |
Chr.7:45806637-45814316 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyth2 | NM_001112701.1 | NP_001106171.1 | ||
| NM_011181.3 | NP_035311.1 | |||
| AB013466.1 | BAA33429.1 | |||
| AB013467.1 | BAA33430.1 | |||
| AF079971.1 | AAC77924.1 | |||
| AK045451.1 | BAC32376.1 | |||
| AK078067.1 | ||||
| AK143829.1 | BAE25558.1 | |||
| AK168979.1 | BAE40780.1 | |||
| AK172050.1 | BAE42799.1 | |||
| BC004662.1 | AAH04662.1 | |||
| BC088833.1 | AAH88833.1 |
More Information
Set Membership: |
|

Genomic Map