Assay Details
Target Gene Details
Entrez Gene ID: | 19158 |
Gene Name: | cytohesin 2 |
Gene Aliases: |
ARNO, CLM2, Pscd2 |
Location: |
Chr.7:45806637-45814316 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyth2 | NM_001112701.1 | 4 | 489 | NP_001106171.1 |
| NM_011181.3 | 4 | 489 | NP_035311.1 | |
| AB013466.1 | 4 | 424 | BAA33429.1 | |
| AB013467.1 | 3 | 992 | BAA33430.1 | |
| AF079971.1 | 4 | 458 | AAC77924.1 | |
| AK045451.1 | 4 | 458 | BAC32376.1 | |
| AK078067.1 | 4 | 485 | ||
| AK143829.1 | 4 | 431 | BAE25558.1 | |
| AK168979.1 | 4 | 429 | BAE40780.1 | |
| AK172050.1 | 4 | 445 | BAE42799.1 | |
| BC004662.1 | 4 | 403 | AAH04662.1 | |
| BC088833.1 | 4 | 463 | AAH88833.1 |
More Information
Set Membership: |
|

Genomic Map