Assay Details
Target Gene Details
Entrez Gene ID: | 68952 |
Gene Name: | family with sequence similarity 57, member B |
Gene Aliases: |
1500016O10Rik, A330104J06Rik, AI413816, AW060769 |
Location: |
Chr.7:126812688-126830219 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam57b | NM_026884.1 | NP_081160.1 | ||
| AK020734.1 |

Genomic Map