Assay Details
Target Gene Details
Entrez Gene ID: | 15388 |
Gene Name: | heterogeneous nuclear ribonucleoprotein L |
Gene Aliases: |
C79783, D830027H13Rik, Hnrpl |
Location: |
Chr.7:28808546-28822268 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpl | NM_177301.5 | NP_796275.3 | ||
| AK044748.1 | ||||
| AK085906.1 | BAC39565.1 | |||
| AK144684.1 | BAE26011.1 | |||
| BC027206.1 | AAH27206.1 | |||
| BC099683.1 | AAH99683.1 |

Genomic Map