Assay Details
Target Gene Details
Entrez Gene ID: | 80719 |
Gene Name: | immunoglobulin superfamily, member 6 |
Gene Aliases: |
- |
Location: |
Chr.7:121064067-121074903 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf6 | NM_030691.1 | NP_109616.1 | ||
| AJ278575.1 | CAC20697.1 | |||
| AK040569.1 | BAC30630.1 | |||
| AK151417.1 | BAE30382.1 | |||
| AK152186.1 | BAE31017.1 | |||
| AK156382.1 | BAE33693.1 | |||
| AK163058.1 | ||||
| BC057951.1 | AAH57951.1 |
Target Gene Details
Entrez Gene ID: | 59052 |
Gene Name: | methyltransferase like 9 |
Gene Aliases: |
0610012D09Rik, AA517660, Drev, MNCb-5680 |
Location: |
Chr.7:121032794-121076835 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl9 | NM_021554.2 | NP_067529.2 | ||
| AB041664.1 | BAA95109.1 | |||
| AJ278576.1 | CAC20698.1 | |||
| AK075586.1 | ||||
| AK169951.1 | BAE41478.1 | |||
| AK188768.1 | ||||
| AK206681.1 | ||||
| AK216652.1 | ||||
| AK216736.1 | ||||
| AK219043.1 | ||||
| BC023188.1 | AAH23188.1 | |||
| BC068124.1 | AAH68124.1 | |||
| BC082317.1 | AAH82317.1 |
More Information
Set Membership: |
|

Genomic Map