Assay Details
Target Gene Details
Entrez Gene ID: | 16068 |
Gene Name: | interleukin 18 binding protein |
Gene Aliases: |
IL-18BP, Igifbp, MC54L |
Location: |
Chr.7:102015077-102019041 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il18bp | NM_010531.1 | NP_034661.1 | ||
| AF110802.1 | 1 | 1064 | AAD17193.1 | |
| AF110803.1 | AAD17194.1 | |||
| AF122907.1 | AAD41052.1 | |||
| AK003370.1 | BAB22744.1 | |||
| AK008452.1 | BAB25677.1 | |||
| AK009721.1 | BAB26462.1 | |||
| BC018332.1 | AAH18332.1 |

Genomic Map