Assay Details
Target Gene Details
Entrez Gene ID: | 53320 |
Gene Name: | folate hydrolase 1 |
Gene Aliases: |
GCP2, mopsm |
Location: |
Chr.7:86718781-86776055 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Folh1 | NM_001159706.1 | NP_001153178.1 | ||
| NM_016770.3 | NP_058050.3 | |||
| AF026380.1 | AAB81971.1 | |||
| AK002920.1 | BAB22457.1 | |||
| BC119604.1 | ||||
| BC119605.1 |
More Information
Set Membership: |
|

Genomic Map