Assay Details
Target Gene Details
Entrez Gene ID: | 14447 |
Gene Name: | glyceraldehyde-3-phosphate dehydrogenase, spermatogenic |
Gene Aliases: |
Gapd-s, Gapds |
Location: |
Chr.7:30729777-30744791 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gapdhs | NM_001290631.1 | NP_001277560.1 | ||
| NM_008085.2 | NP_032111.1 | |||
| AK131805.1 | BAE20812.1 | |||
| M60978.1 | AAA53033.1 |
More Information
Set Membership: |
|

Genomic Map