Assay Details
Target Gene Details
Entrez Gene ID: | 14275 |
Gene Name: | folate receptor 1 (adult) |
Gene Aliases: |
FBP1, Folbp-1, Folbp1 |
Location: |
Chr.7:101858331-101870788 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Folr1 | NM_001252552.1 | 4 | 567 | NP_001239481.1 |
| NM_001252553.1 | 4 | 493 | NP_001239482.1 | |
| NM_001252554.1 | 4 | 480 | NP_001239483.1 | |
| NM_008034.3 | 4 | 564 | NP_032060.2 | |
| AF096319.1 | 4 | 459 | AAD19353.1 | |
| AF096320.1 | 2 | 354 | AAD19354.1 | |
| AK002275.1 | 4 | 562 | ||
| BC023477.1 | 1 | 114 | ||
| BC037078.1 | 3 | 305 | AAH37078.1 | |
| BC138781.1 | 3 | 197 | ||
| BC138795.1 | 3 | 284 | ||
| BC145414.1 | 3 | 284 | ||
| CD241494.1 | 4 | 577 | ||
| CF617679.1 | 3 | 310 | ||
| M64782.1 | 1 | 133 | AAA37595.1 |

Genomic Map