Assay Details
Target Gene Details
Entrez Gene ID: | 14276 |
Gene Name: | folate receptor 2 (fetal) |
Gene Aliases: |
FBP2, FR-P3, FR-beta, Folbp-2, Folbp2 |
Location: |
Chr.7:101839988-101857190 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Folr2 | NM_008035.2 | NP_032061.1 | ||
| BC022108.1 | AAH22108.1 | |||
| BQ176722.1 | ||||
| M64817.1 | AAA37599.1 |

Genomic Map