Assay Details
Target Gene Details
Entrez Gene ID: | 100040049 |
Gene Name: | RIKEN cDNA 1810010D01 gene |
Gene Aliases: |
Gm2572 |
Location: |
Chr.7:145205818-145208119 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810010D01Rik | NR_033626.1 | |||
| AK007416.1 |
Target Gene Details
Entrez Gene ID: | 233979 |
Gene Name: | two pore segment channel 2 |
Gene Aliases: |
BC025890, D830047E22Rik |
Location: |
Chr.7:145181633-145283987 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpcn2 | AK090059.1 | BAC41072.1 |
More Information
Set Membership: |
|

Genomic Map