Assay Details
Target Gene Details
Entrez Gene ID: | 109815 |
Gene Name: | VCP-interacting membrane protein |
Gene Aliases: |
1500011E07Rik, C78786, H-4, H-47, H4, H47, Sels |
Location: |
Chr.7:66079649-66089405 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vimp | NM_024439.3 | NP_077759.3 | ||
| AF335543.1 | AAK07659.1 | |||
| AK005204.1 | BAB23882.1 | |||
| BC011091.1 | AAH11091.1 | |||
| BY335100.1 |

Genomic Map