Assay Details
Target Gene Details
Entrez Gene ID: | 233908 |
Gene Name: | fused in sarcoma |
Gene Aliases: |
D430004D17Rik, D930039C12Rik, Fus1, Tls |
Location: |
Chr.7:127966788-127982032 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fus | NM_139149.2 | NP_631888.1 | ||
| AF224264.1 | AAF70602.1 | |||
| AK032815.1 | ||||
| AK038226.1 | ||||
| AK134491.1 | ||||
| AK139960.1 | BAE24196.1 | |||
| AK145582.1 | BAE26524.1 | |||
| AK146198.1 | BAE26972.1 | |||
| AK149118.1 | ||||
| AK172554.1 | ||||
| BC011078.1 | AAH11078.1 | |||
| BC040827.1 | AAH40827.1 | |||
| BC058247.1 | AAH58247.1 | |||
| BY269119.1 |
More Information
Set Membership: |
|

Genomic Map